Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caveolin-1 Antibody (7C8), Novus Biologicals™
SDP

Catalog No. NB100615 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100615 0.1 mL
NB100615SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100615 Supplier Novus Biologicals Supplier No. NB100615
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 25 publications

Caveolin-1 Monoclonal specifically detects Caveolin-1 in Drosophila melanogaster, Human, Mouse, Ovine, Rat, Sheep samples. It is validated for Flow Cytometry, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Caveolin-1
Applications Flow Cytometry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 7C8
Concentration 1.25 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:4000, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:100-1:300, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1-2 ug/ 500 ug of protein, Immunohistochemistry-Paraffin 1:100-1:300, Immunohistochemistry-Frozen reported in scientific literature (PMID 24758774), Immunoblotting reported in scientific literature (PMID 31759628)
Gene Alias CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085
Gene Symbols CAV1
Host Species Mouse
Immunogen Intracellular membrane protein-containing vesicles (containing GLUT4) from rat adipocytes
Molecular Weight of Antigen 23 kDa
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Caveolae Markers, Cellular Markers, Cholesterol Metabolism, Hypoxia, Lipid and Metabolism, Lipid Droplets, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 857
Test Specificity This is specific for caveolin alpha and beta proteins.
Target Species Drosophila melanogaster, Mouse, Human, Mouse, Ovine, Rat, Sheep
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.