Learn More
Description
Specifications
Specifications
| Antigen | Caveolin-1 |
| Applications | Flow Cytometry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 7C8 |
| Concentration | 1.25 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:4000, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:100-1:300, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1-2 ug/ 500 ug of protein, Immunohistochemistry-Paraffin 1:100-1:300, Immunohistochemistry-Frozen reported in scientific literature (PMID 24758774), Immunoblotting reported in scientific literature (PMID 31759628) |
| Gene Alias | CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085 |
| Gene Symbols | CAV1 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
