Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caveolin-1 Antibody (7C8), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB10615A594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10615A594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10615A594 Supplier Novus Biologicals Supplier No. NB100615AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Caveolin-1 Monoclonal specifically detects Caveolin-1 in Human, Mouse, Rat, Sheep samples. It is validated for Flow Cytometry, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Caveolin-1
Applications Flow Cytometry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 7C8
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085
Gene Symbols CAV1
Host Species Mouse
Immunogen Intracellular membrane protein-containing vesicles (containing GLUT4) from rat adipocytes
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 857
Test Specificity This is specific for caveolin alpha and beta proteins.
Target Species Human, Mouse, Rat, Sheep
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.