Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caveolin-1 Antibody (7C8), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB303223 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303223 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303223 Supplier Novus Biologicals Supplier No. FAB5736D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Caveolin-1 Monoclonal antibody specifically detects Caveolin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry

Specifications

Antigen Caveolin-1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7C8
Conjugate DyLight 350
Dilution Western Blot, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias CAV, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, cell growth-inhibiting protein 32, MSTP085
Host Species Mouse
Immunogen Puified rat adipocyte low density microsomes, Accession # P41350
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Caveolae Markers, Cellular Markers, Cholesterol Metabolism, Hypoxia, Lipid and Metabolism, Lipid Droplets, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 857
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.