Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caveolin-3 Antibody, Novus Biologicals™
SDP

Catalog No. NB110502902 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB110502902 0.02 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB110502902 Supplier Novus Biologicals Supplier No. NB11050290.02MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Caveolin-3 Polyclonal specifically detects Caveolin-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Caveolin-3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Flow Cytometry 3 - 5 ug/ 10^6 cells, Immunohistochemistry 1:100 - 200, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunoprecipitation 5 ug, Immunohistochemistry-Paraffin 1:100 - 200
Formulation PBS, 1 mg/ml BSA with 0.05% Sodium Azide
Gene Accession No. P56539
Gene Alias caveolin 3, caveolin-3, LGMD1CMGC126100, LQT9MGC126101, M-caveolin, MGC126129, VIP21, VIP-21
Gene Symbols CAV3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues M(1) M T E E H T D L E A R I I K D I H C(19) C of mouse CAV3
Purification Method Affinity Purified
Quantity 0.02 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 859
Test Specificity Detects caveolin-3 from rat tissues. This does not detect caveolin-1 or -2.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.