Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caveolin-3 Rabbit anti-Human, Clone: SY22-06, Novus Biologicals™
SDP

Catalog No. NBP267424 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267424 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267424 Supplier Novus Biologicals Supplier No. NBP267424

Rabbit Monoclonal Antibody

Caveolin-3 Monoclonal specifically detects Caveolin-3 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Caveolin-3
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone SY22-06
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Immunoprecipitation
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias caveolin 3, caveolin-3, LGMD1CMGC126100, LQT9MGC126101, M-caveolin, MGC126129, VIP21, VIP-21
Gene Symbols CAV3
Host Species Rabbit
Immunogen Recombinant protein within Human Caveolin-3 aa 1-125/ 151. (SwissProt: P56539 Human; SwissProt: P51637 Mouse; SwissProt: P51638 Rat)
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 859
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.