Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cbl-c Antibody (OTI3B4), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB278509 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB278509 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB278509 Supplier Novus Biologicals Supplier No. NBP271755MFV610

Mouse Monoclonal Antibody

Cbl-c Monoclonal antibody specifically detects Cbl-c in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Cbl-c
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3B4
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias Cas-Br-M (murine) ecotropic retroviral transforming sequence c, Cas-Br-M (murine) ectropic retroviral transforming sequence c, CBL3, CBL-3, CBL-SL, RING finger protein 57, RNF57SH3-binding protein CBL-3, SH3-binding protein CBL-C, signal transduction protein CBL-C
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 244-474 of human CBLC(NP_036248) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 23624
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.