Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cbl-c Antibody (OTI4E3), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB271756A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271756A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271756A48 Supplier Novus Biologicals Supplier No. NBP271756AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cbl-c Monoclonal specifically detects Cbl-c in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen Cbl-c
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI4E3
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cas-Br-M (murine) ecotropic retroviral transforming sequence c, Cas-Br-M (murine) ectropic retroviral transforming sequence c, CBL3, CBL-3, CBL-SL, RING finger protein 57, RNF57SH3-binding protein CBL-3, SH3-binding protein CBL-C, signal transduction protein CBL-C
Gene Symbols CBLC
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 244-474 of human Cbl-c (NP_036248) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 23624.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.