Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBP/KAT3A Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB055198 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB055198 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB055198 Supplier Novus Biologicals Supplier No. NBP298753AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CBP/KAT3A Polyclonal specifically detects CBP/KAT3A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CBP/KAT3A
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CBPRTS, CREB binding protein, CREB-binding protein, EC 2.3.1.48, KAT3A, RSTS, Rubinstein-Taybi syndrome
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human CBP/KAT3A.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Chromatin Research, HIF Target Genes, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1387
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.