Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBP/KAT3A Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB363098 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363098 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363098 Supplier Novus Biologicals Supplier No. NB100381DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CBP/KAT3A Polyclonal antibody specifically detects CBP/KAT3A in Human samples. It is validated for Western Blot, Immunoprecipitation, ChIP assay (ChIP)

Specifications

Antigen CBP/KAT3A
Applications Western Blot, Immunoprecipitation, ChIP Assay
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP)
Formulation 50mM Sodium Borate
Gene Alias CBPRTS, CREB binding protein, CREB-binding protein, EC 2.3.1.48, KAT3A, RSTS, Rubinstein-Taybi syndrome
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 975 and 1025 of human CREB Binding Protein using the numbering given in entry NP_004371.1 (GeneID 1387).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Chromatin Research, HIF Target Genes, Hypoxia, Membrane Trafficking and Chaperones, Signal Transduction, Stem Cell Signaling Pathway, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1387
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.