Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBP20 Antibody, Novus Biologicals™
SDP

Catalog No. NBP130445 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130445 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130445 Supplier Novus Biologicals Supplier No. NBP130445
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CBP20 Polyclonal specifically detects CBP20 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen CBP20
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry, Immunoprecipitation 5-15 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. P52298
Gene Alias 20 kDa nuclear cap-binding protein, Cbc2, CBP20CBC2, NCBP 20 kDa subunit, NCBP interacting protein 1, NCBP-interacting protein 1, NIP1Cell proliferation-inducing gene 55 protein, nuclear cap binding protein subunit 2, 20kD, nuclear cap binding protein subunit 2, 20kDa, nuclear cap-binding protein subunit 2
Gene Symbols NCBP2
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 106 to 156 of human nuclear cap binding protein subunit 2 using the numbering given in entry NP_031388.2 (GeneID 22916).
Molecular Weight of Antigen 18 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 22916
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.