Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBR3 Antibody (OTI1G6), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP270343V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270343V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270343V Supplier Novus Biologicals Supplier No. NBP270343V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CBR3 Monoclonal specifically detects CBR3 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen CBR3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1G6
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias carbonyl reductase (NADPH) 3, carbonyl reductase (NADPH) 3, EC 1.1.1.18410EC 1.1.1, carbonyl reductase [NADPH] 3, carbonyl reductase 3, EC 1.1.1.184, hCBR3, NADPH-dependent carbonyl reductase 3, SDR21C2, short chain dehydrogenase/reductase family 21C, member 2
Gene Symbols CBR3
Host Species Mouse
Immunogen Full length recombinant protein of human CBR3 (NP_001227) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 874.0
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.