Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBS Scientific Standard Submarine Horizontal Electrophoresis Systems
SCI_ED

Catalog No. p-6666071
Click to view available options
Gel Dimensions:
20 x 20 cm
20 x 30 cm
20 x 40 cm

Four sizes with three options for gel casting.

CBS Economy Horizontal Productivity Packages feature platinum electrode cassettes which provide rapid sample velocity and clear focused bands without smiling and cab be easily removed for cleaning or replacement.

Gel Tray Specifications:

  • Winged gel trays for easy handling - no more sticking fingers into slippery buffer to extract gel tray
  • Made from sturdy 6.4mm thick UV transparent acrylic
  • Grooves in gel tray corners create agarose anchors to prevent gel flotation during electrophoresis
  • Gel tray configuration allows single run or multiple simultaneous short runs
  • Three options for gel casting; tapered baffles, External Gel Casters and Continuously Adjustable Caster
  • Adjustable height polycarbonate or PTFE-coated aluminum combs are available in many configurations, with or without markers
  • PTFE-coated combs allow faster well formation and no wells are lost when using low percentage or low melt agarose
  • Buffer recirculation ports allow maintenance of uniform pH and ionic strength and reduce frequency of buffer changes necessary during long runs
  • Tight-fitting safety cover reduces condensation during electrophoresis
  • Coolant can be recirculated beneath gel bed to provide constant temperature runs
  • Combs are printed with catalog numbers and thickness for easy identification

Applications:
Seperation and identification of DNA fragments, RNA fragments, PCR Products and synthetic oligonucleotides

Specifications

Certifications/Compliance CE
Color Blue
Description Compatible Gel Tray: WGT-020
Type Horizontal Gel Electrophoresis System
Gel Dimensions 20 x 20 cm
Volume (Metric) Buffer 1850 mL
Warranty 3 Years
Dimensions (L x W) Gel Tray I.D.: 18.7 x 20 cm (7.3 x 7.8 in.)
Dimensions (L x W x H) 43 x 21 x 10 cm (16.9 x 8.26 x 3.9 in.)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.