Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cbx2 Antibody (RAB-C238), PerCP, Novus Biologicals™
SDP

Catalog No. NB340462 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340462 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340462 Supplier Novus Biologicals Supplier No. NBP320135PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cbx2 Monoclonal antibody specifically detects Cbx2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, ChIP assay (ChIP)

Specifications

Antigen Cbx2
Applications Flow Cytometry, ELISA, Immunocytochemistry, Immunoprecipitation, Mass Spectrometry
Classification Monoclonal
Clone RAB-C238
Conjugate PerCP
Dilution Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Mass Spectrometry, Chromatin Immunoprecipitation (ChIP)
Formulation PBS
Gene Alias CDCA6, cell division cycle associated 6, chromobox homolog 2, chromobox homolog 2 (Drosophila Pc class), chromobox homolog 2 (Pc class homolog, Drosophila), chromobox protein homolog 2, M33, MGC10561, modifier 3, Pc class homolog
Host Species Rabbit
Immunogen This antibody was obtained by recombinant antibody (rAb) phage display recognizing CBX2 protein under non-denaturing conditions.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Modifiers
Primary or Secondary Primary
Gene ID (Entrez) 84733
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.