Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBX3 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB105588 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB105588 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB105588 Supplier Novus Biologicals Supplier No. NBP298757DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CBX3 Polyclonal specifically detects CBX3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CBX3
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias chromobox homolog 3, chromobox homolog 3 (Drosophila HP1 gamma), chromobox protein homolog 3, Drosophila), HECH, Heterochromatin protein 1 homolog gamma, heterochromatin protein HP1 gamma, heterochromatin-like protein 1, HP1 gamma, HP1 gamma homolog, HP1-GAMMA, Modifier 2 protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human CBX3.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 11335
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.