Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CC Chemokine Receptor D6 Antibody
SDP

Catalog No. NB017796 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017796 0.025 mg
NB100703 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017796 Supplier Novus Biologicals Supplier No. NB1007030.025MG

Goat Polyclonal Antibody

CC Chemokine Receptor D6 Polyclonal specifically detects CC Chemokine Receptor D6 in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CC Chemokine Receptor D6
Applications Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry, Flow Cytometry
Classification Polyclonal
Concentration 2.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1-5ul/ml, ELISA 1:50000, Immunohistochemistry 1:250, Immunohistochemistry-Paraffin 1:250
Formulation 10 mM KHPO4, 140 mM NaCl, BSA @ 1mg/ml with 0.1% Sodium Azide
Gene Alias C-C chemokine receptor D6, CCR10CCR9D6CC-chemokine-binding receptor JAB61, chemokine (C-C) receptor 9, chemokine binding protein 2, Chemokine receptor CCR-10, Chemokine receptor CCR-9, chemokine receptor D6, chemokine-binding protein 2, Chemokine-binding protein D6, CMKBR9chemokine (C-C motif) receptor 9, hD6, MGC126678, MGC138250
Gene Symbols CCBP2
Host Species Goat
Immunogen Synthetic peptide SSSIYDYDYLDDMTILVCRKDEVLSFGR corresponding to aa 18-45 of mouse beta chemokine receptor D6
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1238
Test Specificity Peptide sequence is < 50 % identical to other mouse chemokine receptors in this region.
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.