Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CC Chemokine Receptor D6 Antibody (196124), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB175149 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175149 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175149 Supplier Novus Biologicals Supplier No. FAB1364Z
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CC Chemokine Receptor D6 Monoclonal antibody specifically detects CC Chemokine Receptor D6 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CC Chemokine Receptor D6
Applications Flow Cytometry
Classification Monoclonal
Clone 196124
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias C-C chemokine receptor D6, CCR10CCR9D6CC-chemokine-binding receptor JAB61, chemokine (C-C) receptor 9, chemokine binding protein 2, Chemokine receptor CCR-10, Chemokine receptor CCR-9, chemokine receptor D6, chemokine-binding protein 2, Chemokine-binding protein D6, CMKBR9chemokine (C-C motif) receptor 9, hD6, MGC126678, MGC138250
Host Species Rat
Immunogen RBL-1 rat basophilic leukemia cell line transfected with human CC Chemokine Receptor D6, Met1-Ala384, Accession # O00590
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Autophagy, Cellular Markers, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1238
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.