Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCBL2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157482
Description
CCBL2 Polyclonal specifically detects CCBL2 in Human samples. It is validated for Western Blot.Specifications
CCBL2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
CCBL2 | |
Synthetic peptides corresponding to CCBL2(cysteine conjugate-beta lyase 2) The peptide sequence was selected from the C terminal of CCBL2. Peptide sequence LSAIPVSAFCNSETKSQFEKFVRFCFIKKDSTLDAAEEIIKAWSVQKS. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Canine: 100%; Guinea pig: 100%; Human: 100%; Equine: 92%; Pig: 92%; Chicken: 85%; Rabbit: 85%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
cysteine conjugate-beta lyase 2, Cysteine-S-conjugate beta-lyase 2, DKFZp547N1117, EC 2.6.1.63, EC 2.6.1.7, EC 4.4.1.13, KAT3DKFZp667D0223, KATIII, Kynurenine aminotransferase III, Kynurenine--glyoxylate transaminase, kynurenine-oxoglutarate transaminase 3, kynurenine--oxoglutarate transaminase 3, Kynurenine--oxoglutarate transaminase III, MGC9398, RBM1, RP11-82K18.3, RP4-531M19.2 | |
Rabbit | |
Affinity purified | |
RUO | |
56267 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction