Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCBL2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | CCBL2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CCBL2 Polyclonal specifically detects CCBL2 in Human samples. It is validated for Western Blot.Specifications
CCBL2 | |
Polyclonal | |
Rabbit | |
cysteine conjugate-beta lyase 2, Cysteine-S-conjugate beta-lyase 2, DKFZp547N1117, EC 2.6.1.63, EC 2.6.1.7, EC 4.4.1.13, KAT3DKFZp667D0223, KATIII, Kynurenine aminotransferase III, Kynurenine--glyoxylate transaminase, kynurenine-oxoglutarate transaminase 3, kynurenine--oxoglutarate transaminase 3, Kynurenine--oxoglutarate transaminase III, MGC9398, RBM1, RP11-82K18.3, RP4-531M19.2 | |
CCBL2 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
56267 | |
Synthetic peptides corresponding to CCBL2(cysteine conjugate-beta lyase 2) The peptide sequence was selected from the C terminal of CCBL2. Peptide sequence LSAIPVSAFCNSETKSQFEKFVRFCFIKKDSTLDAAEEIIKAWSVQKS. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title