Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC112 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159468
Description
CCDC112 Polyclonal specifically detects CCDC112 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CCDC112 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
coiled-coil domain containing 112, coiled-coil domain-containing protein 112, MBC1, MGC39633, mutated in bladder cancer 1, Mutated in bladder cancer protein 1 | |
Rabbit | |
Protein A purified | |
RUO | |
153733 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Q6A334 | |
CCDC112 | |
Synthetic peptides corresponding to MGC39633 The peptide sequence was selected from the N terminal of MGC39633. Peptide sequence VRTAEKFKNQVINMEKDKHSHFYNQKSDFRIEHSMLEELENKLIHSRKTE. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Crab-eating macaque: 100%; Mouse: 85%; Rat: 78%;. | |
Human, Mouse, Rat, Pig, Bovine, Equine, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction