Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC141 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB143091 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB143091 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB143091 Supplier Novus Biologicals Supplier No. NBP305861F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC141 Polyclonal specifically detects CCDC141 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CCDC141
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias coiled-coil domain containing 141, DKFZp686G197, MGC134803
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CCDC141 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285025
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.