Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC16 Antibody, Novus Biologicals™
SDP

Catalog No. NB10068229 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10068229 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10068229 Supplier Novus Biologicals Supplier No. NB10068229
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC16 Polyclonal specifically detects CCDC16 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen CCDC16
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_443089.3
Gene Alias CCDC16, coiled-coil domain containing 16, Coiled-coil domain-containing protein 16, MGC20398, OMCG1, SEL13, sel-13 suppressor of lin-12 homolog, zinc finger protein 830
Gene Symbols ZNF830
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 322 and 372 of human coiled-coil domain containing 16 using the numbering given in entry NP_443089.3 (GeneID 91603).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91603
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Panda, Orangutan, Rhesus Monkey, Chimpanzee, Crabeatingmacaque, African elephant, Naked mole rat and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.