Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC44 Antibody, Novus Biologicals™
SDP

Catalog No. NBP215750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215750 Supplier Novus Biologicals Supplier No. NBP215750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC44 Polyclonal specifically detects CCDC44 in Human samples. It is validated for Western Blot.

Specifications

Antigen CCDC44
Applications Western Blot
Classification Polyclonal
Concentration 0.59mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9BSH4
Gene Alias CCDC44translational activator of cytochrome c oxidase 1, clone HQ0477 PRO0477p, coiled-coil domain containing 44, Coiled-coil domain-containing protein 44, translational activator of mitochondrially encoded cytochrome c oxidase I, Translational activator of mitochondrially-encoded cytochrome c oxidase I
Gene Symbols TACO1
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human CCDC44. The exact sequence is proprietary.
Molecular Weight of Antigen 32 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51204
Test Specificity Human. Expected to cross react based on sequence identity: Monkey (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.