Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC47 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB184768 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184768 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184768 Supplier Novus Biologicals Supplier No. NBP282077JF646
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CCDC47 Polyclonal antibody specifically detects CCDC47 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CCDC47
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias coiled-coil domain containing 47, coiled-coil domain-containing protein 47, GK001, MSTP041, PSEC0077
Host Species Rabbit
Immunogen CCDC47 antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human CCDC47. The immunogen is located within the last 50 amino acids of CCDC47. Amino Acid Squence: RREQKKLEKKQMKMK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 57003
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.