Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC5 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB180461 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB180461 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB180461 Supplier Novus Biologicals Supplier No. NBP306419H
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CCDC5 Polyclonal antibody specifically detects CCDC5 in Human samples. It is validated for Western Blot

Specifications

Antigen CCDC5
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias CCDC5Enhancer of invasion-cluster, coiled-coil domain containing 5 (spindle associated), FLJ40084, HAUS augmin-like complex subunit 1, HAUS augmin-like complex, subunit 1, HEI-CCoiled-coil domain-containing protein 5, HEICFLJ21094, HsT1461
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CCDC5 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 115106
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.