Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC60 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15674720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15674720 20 μL
NBP156747 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15674720 Supplier Novus Biologicals Supplier No. NBP15674720UL

Rabbit Polyclonal Antibody

CCDC60 Polyclonal specifically detects CCDC60 in Human samples. It is validated for Western Blot.

Specifications

Antigen CCDC60
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8IWA6
Gene Alias coiled-coil domain containing 60, coiled-coil domain-containing protein 60, MGC39827
Gene Symbols CCDC60
Host Species Rabbit
Immunogen Synthetic peptides corresponding to CCDC60(coiled-coil domain containing 60) The peptide sequence was selected from the C terminal of CCDC60. Peptide sequence RPAKKILVKLQKFGENLDLRIRPHVLLKVLQDLRIWELCSPDIAVAIEFV.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 160777
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.