Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC60 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5122148
This item is not returnable.
View return policy
Description
CCDC60 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
CCDC60 is a protein coding gene.
Specifications
CCDC60 | |
Polyclonal | |
Unconjugated | |
CCDC60 | |
AU018954; C130098D09; Ccdc60; coiled-coil domain containing 60; coiled-coil domain-containing protein 60; LOW QUALITY PROTEIN: coiled-coil domain-containing protein 60 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
269693 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q8C4J0 | |
CCDC60 | |
Recombinant protein Coiled Coil Domain Containing Protein 60. The antigen corresponds to amino acid range 1-186 of the target protein. | |
20 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction