Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CCDC60 Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $577.00
Specifications
Antigen | CCDC60 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119038
|
Invitrogen™
PA5119038 |
200 μL |
Each for $577.00
|
|
|||||
PIPA5122148
|
Invitrogen™
PA5122148 |
20 μL |
Each for $237.00
|
|
|||||
Description
CCDC60 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
CCDC60 is a protein coding gene.Specifications
CCDC60 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Mouse | |
CCDC60 | |
AU018954; C130098D09; Ccdc60; coiled-coil domain containing 60; coiled-coil domain-containing protein 60; LOW QUALITY PROTEIN: coiled-coil domain-containing protein 60 | |
CCDC60 | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q8C4J0 | |
269693 | |
Recombinant protein Coiled Coil Domain Containing Protein 60. The antigen corresponds to amino acid range 1-186 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title