Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC66 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB290884 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290884 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290884 Supplier Novus Biologicals Supplier No. NBP178739PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC66 Polyclonal antibody specifically detects CCDC66 in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative)

Specifications

Antigen CCDC66
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation (Negative)
Formulation PBS
Gene Alias coiled-coil domain containing 66, coiled-coil domain-containing protein 66, DKFZp686C0433
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 898 and 948 of human Coiled-Coil Domain Containing 66 using the numbering given in entry NP_001135419.1 (GeneID 285331).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285331
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.