Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC7 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$208.00 - $487.50

Specifications

Antigen CCDC7
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15675720
SDP
View Documents
Novus Biologicals
NBP15675720UL
20 μL
Each for $208.00
Only null left
Add to Cart
 
NBP156757
SDP
View Documents
Novus Biologicals
NBP156757
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

CCDC7 Polyclonal specifically detects CCDC7 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CCDC7
Polyclonal
Rabbit
Q96M83
221016
Synthetic peptides corresponding to CCDC7(coiled-coil domain containing 7) The peptide sequence was selected from the N terminal of CCDC7. Peptide sequence KHNAKLIHDKIEPMVLRSPPTGESILRYALPIPSSKTKNLLPEDEMIGKI.
Primary
Western Blot
Unconjugated
RUO
BioT2-A, BioT2-B, BioT2-C, coiled-coil domain containing 7, coiled-coil domain-containing protein 7, DKFZp686N0559, FLJ32762, RP11-479G22.1
CCDC7
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.