Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCDC74A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP156895
Description
CCDC74A Polyclonal specifically detects CCDC74A in Human samples. It is validated for Western Blot.Specifications
| CCDC74A | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| coiled-coil domain containing 74A, coiled-coil domain-containing protein 74A, FLJ40345 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 90557 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q96AQ1 | |
| CCDC74A | |
| Synthetic peptides corresponding to CCDC74A(coiled-coil domain containing 74A) The peptide sequence was selected from the middle region of CCDC74A. Peptide sequence FPKVSTKSLSKKCLSPPVAERAILPALKQTPKNNFAERQKRLQAMQKRRL. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Mouse: 85%. | |
| Human, Mouse, Equine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction