Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCDC9 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB143589 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB143589 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB143589 Supplier Novus Biologicals Supplier No. NBP305897V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCDC9 Polyclonal specifically detects CCDC9 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CCDC9
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias coiled-coil domain containing 9, coiled-coil domain-containing protein 9, DKFZP586M1019
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CCDC9 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26093
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.