Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCK-AR Antibody (377251), PerCP, Novus Biologicals™
SDP

Catalog No. NB364505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364505 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364505 Supplier Novus Biologicals Supplier No. FAB2680C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCK-AR Monoclonal antibody specifically detects CCK-AR in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen CCK-AR
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 377251
Conjugate PerCP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias CCK1-R, CCK-A, CCK-A receptor, CCK-AR, CCKRA, cholecystokinin A receptor, cholecystokinin receptor type A, cholecystokinin type-A receptor, Cholecystokinin-1 receptor
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CCK-A R, Met1-Gln428, Accession # P32238
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, GPCR, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 886
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.