Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CCK Polyclonal Antibody

Catalog No. p-7168036
Change view
Click to view available options
Quantity:
500 μL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-673-190 1 mL
PIPA532348 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-673-190 Supplier Invitrogen™ Supplier No. PA137267
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is stomach tissue.

Cholecystokinin (CCK) is a neuropeptide hormone and neurotransmitter widely distributed throughout the gastrointestinal (GI) tract and the central nervous system (CNS). Antibodies that react specifically with CCK are useful for the study of the mode of action, differential tissue expression and intracellular and subcellular localization of CCK in the CNS and periphery as well as in neuroendocrine cells of the digestive system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CCK
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.1% sodium azide; pH 7.6
Gene CCK
Gene Accession No. P06307
Gene Alias (1-49)-CCK58; CCK; CCK12; CCK18; CCK22; CCK25; CCK33; CCK39; CCK5; CCK58; CCK7; CCK8; Cholecystokinin; cholecystokinin octapeptide; cholecystokinin triacontatriapeptide; Cholecystokinin-12; Cholecystokinin-18; Cholecystokinin-22; Cholecystokinin-25; Cholecystokinin-33; Cholecystokinin-39; Cholecystokinin-5; Cholecystokinin-58; Cholecystokinin-58 desnonopeptide; Cholecystokinin-7; Cholecystokinin-8; prepro-cholecystokinin
Gene Symbols CCK
Host Species Rabbit
Immunogen Synthetic peptide within Human Cholecystokinin aa 50 to the C-terminus (N terminal).
Purification Method Antigen affinity chromatography
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 885
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.