Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL1/I-309/TCA-3 Antibody (RM0167-6H50), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP211876A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP211876A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP211876A94 Supplier Novus Biologicals Supplier No. NBP211876AF594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CCL1/I-309/TCA-3 Monoclonal specifically detects CCL1/I-309/TCA-3 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen CCL1/I-309/TCA-3
Applications Western Blot
Classification Monoclonal
Clone RM0167-6H50
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C-C motif chemokine 1, chemokine (C-C motif) ligand 1, I-309, inflammatory cytokine I-309, P500, SCYA1Small-inducible cytokine A1, SISe, small inducible cytokine A1 (I-309, homologous to mouse Tca-3), T lymphocyte-secreted protein I-309, TCA3
Gene Symbols CCL1
Host Species Rat
Immunogen Mouse recombinant CCL1/I-309/TCA-3
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6346
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.