Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL11/Eotaxin Antibody (43915), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB304087 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304087 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304087 Supplier Novus Biologicals Supplier No. FAB3201D
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCL11/Eotaxin Monoclonal specifically detects CCL11/Eotaxin in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Western Blot.

Specifications

Antigen CCL11/Eotaxin
Applications CyTOF, Flow Cytometry (Intracellular Staining), Western Blot
Classification Monoclonal
Clone 43915
Conjugate DyLight 350
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias C-C motif chemokine 11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, eotaxin, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin)
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL11/Eotaxin
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Asthma, Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6356
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.