Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL11/Eotaxin Antibody (OTI2H9), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP271277A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP271277A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP271277A70 Supplier Novus Biologicals Supplier No. NBP271277AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCL11/Eotaxin Monoclonal specifically detects CCL11/Eotaxin in Human samples. It is validated for Western Blot.

Specifications

Antigen CCL11/Eotaxin
Applications Western Blot
Classification Monoclonal
Clone OTI2H9
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C-C motif chemokine 11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, eotaxin, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin)
Gene Symbols CCL11
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 24-97 of human CCL11/Eotaxin (NP_002977) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6356
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.