Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL12/MCP-5 Antibody (RM0169-8J25), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB361359 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361359 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361359 Supplier Novus Biologicals Supplier No. NBP212089MFV500

Rat Monoclonal Antibody

CCL12/MCP-5 Monoclonal antibody specifically detects CCL12/MCP-5 in Mouse samples. It is validated for Western Blot, Neutralization

Specifications

Antigen CCL12/MCP-5
Applications Western Blot, Neutralization
Classification Monoclonal
Clone RM0169-8J25
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias C-C motif chemokine 12, chemokine (C-C motif) ligand 12, MCP-5, Scya12
Host Species Rat
Immunogen Mouse recombinant CCL12/MCP-5
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 20293
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.