Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL13/MCP-4 Antibody (73506), PerCP, Novus Biologicals™
SDP

Catalog No. NB296531 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296531 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296531 Supplier Novus Biologicals Supplier No. FAB327C

Mouse Monoclonal Antibody

CCL13/MCP-4 Monoclonal antibody specifically detects CCL13/MCP-4 in Human samples. It is validated for ELISA Standard (Matched Pair), Neutralization, ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair), Flow Cytometry, CyTOF

Specifications

Antigen CCL13/MCP-4
Applications ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Flow Cytometry
Classification Monoclonal
Clone 73506
Conjugate PerCP
Dilution ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias C-C motif chemokine 13, chemokine (C-C motif) ligand 13, CKb10, MCP4, MCP-4Monocyte chemoattractant protein 4, MGC17134, NCC-1Monocyte chemotactic protein 4, NCC1Small-inducible cytokine A13, new CC chemokine 1, SCYA13CK-beta-10, SCYL1, small inducible cytokine subfamily A (Cys-Cys), member 13
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL13/MCP-4, Gln24-Thr98, Accession # Q99616.1
Purification Method Protein A or G purified from ascites
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6357
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.