Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL13/MCP-4 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen CCL13/MCP-4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP17993620
SDP
View Documents
Novus Biologicals
NBP17993620UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP179936
SDP
View Documents
Novus Biologicals
NBP179936
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

CCL13/MCP-4 Polyclonal specifically detects CCL13/MCP-4 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CCL13/MCP-4
Polyclonal
Rabbit
Cancer
NP_005399
6357
Synthetic peptide directed towards the middle region of human CCL13. Peptide sequence KSYVITTSRCPQKAVIFRTKLGKEICADPKEKWVQNYMKHLGRKAHTLKT.
Primary
Western Blot
Unconjugated
RUO
Human
C-C motif chemokine 13, chemokine (C-C motif) ligand 13, CKb10, MCP4, MCP-4Monocyte chemoattractant protein 4, MGC17134, NCC-1Monocyte chemotactic protein 4, NCC1Small-inducible cytokine A13, new CC chemokine 1, SCYA13CK-beta-10, SCYL1, small inducible cytokine subfamily A (Cys-Cys), member 13
CCL13
IgG
11 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.