Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL14/HCC-1/HCC-3 Antibody (015), PE, Novus Biologicals™
SDP

Catalog No. NB307267 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB307267 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB307267 Supplier Novus Biologicals Supplier No. NBP289521PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CCL14/HCC-1/HCC-3 Monoclonal antibody specifically detects CCL14/HCC-1/HCC-3 in Human samples. It is validated for ELISA

Specifications

Antigen CCL14/HCC-1/HCC-3
Applications ELISA
Classification Monoclonal
Clone 015
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias C-C motif chemokine 14, chemokine (C-C motif) ligand 14, Chemokine CC-1/CC-3, chemokine CC-3, CKB1, FLJ16015, HCC-1, HCC-1(1-74), HCC-1/HCC-3, HCC-3, hemofiltrate CC chemokine 1, MCIF, member 14, NCC2CC-1, NCC-2CKb1, new CC chemokine 2, SCYA14CC-3, Small-inducible cytokine A14
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL14/HCC-1/HCC-3 (Uniprot#: Q16627-1; Thr20-Asn93).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6358
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.