Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL15/MIP-1 delta Antibody (001), PerCP, Novus Biologicals™
SDP

Catalog No. NB306515 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306515 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB306515 Supplier Novus Biologicals Supplier No. NBP289703PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CCL15/MIP-1 delta Monoclonal antibody specifically detects CCL15/MIP-1 delta in Human samples. It is validated for ELISA

Specifications

Antigen CCL15/MIP-1 delta
Applications ELISA
Classification Monoclonal
Clone 001
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias chemokine (C-C motif) ligand 15, Chemokine CC-2, HCC-2Lkn-1, leukotactin 1, Leukotactin-1, LKN1, LKN-1, Macrophage inflammatory protein 5, MIP-1 delta, MIP5, MIP-5MIP-1d, Mrp-2b, NCC-3MIP-1D, NCC3MRP-2B, new CC chemokine 3, SCYA15member 15, Small-inducible cytokine A15
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human CCL15/MIP-1 delta (Uniprot#: Q16663-1; Gln22-Ile113).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 6359
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.