Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL16/HCC-4/LEC Antibody (70218), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB304036 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304036 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304036 Supplier Novus Biologicals Supplier No. FAB328V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCL16/HCC-4/LEC Monoclonal specifically detects CCL16/HCC-4/LEC in Human samples. It is validated for ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Neutralization.

Specifications

Antigen CCL16/HCC-4/LEC
Applications ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Neutralization
Classification Monoclonal
Clone 70218
Conjugate Alexa Fluor 405
Dilution ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Formulation 50mM Sodium Borate
Gene Alias C-C motif chemokine 16, chemokine (C-C motif) ligand 16, Chemokine CC-4, Chemokine LEC, CKb12, HCC-4Liver-expressed chemokine, LCC-1Lymphocyte and monocyte chemoattractant, LEC, liver CC chemokine-1, LMCSmall-inducible cytokine A16, monotactin-1, MTN-1, NCC4IL-10-inducible chemokine, NCC-4ILINCK, new CC chemokine 4, SCYA16MGC117051, SCYL4, small inducible cytokine subfamily A (Cys-Cys), member 16
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL16/HCC-4, Gln24-Gln120, Accession # O15467
Purification Method Protein A or G purified from ascites
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6360
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.