Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL17/TARC Antibody (54015), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB303888 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303888 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303888 Supplier Novus Biologicals Supplier No. FAB3641M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCL17/TARC Monoclonal antibody specifically detects CCL17/TARC in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen CCL17/TARC
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 54015
Conjugate DyLight 594
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ABCD-2, CC chemokine TARC, C-C motif chemokine 17, chemokine (C-C motif) ligand 17, MGC138273, SCYA17MGC138271, small inducible cytokine subfamily A (Cys-Cys), member 17, Small-inducible cytokine A17, T cell-directed CC chemokine, TARCA-152E5.3, Thymus and activation-regulated chemokine
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL17/TARC, Ala24-Ser94, Accession # Q92583.1
Purification Method Protein A or G purified from ascites
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Chemokines and Cytokines
Primary or Secondary Primary
Gene ID (Entrez) 6361
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.