Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL18/PARC Antibody (64507), PE, Novus Biologicals™
SDP

Catalog No. NB371122 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371122 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371122 Supplier Novus Biologicals Supplier No. FAB394P
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCL18/PARC Monoclonal specifically detects CCL18/PARC in Human samples. It is validated for CyTOF, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Flow Cytometry (Intracellular Staining), Western Blot.

Specifications

Antigen CCL18/PARC
Applications CyTOF, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), ELISA Standard (Matched Pair), Flow Cytometry (Intracellular Staining), Western Blot
Classification Monoclonal
Clone 64507
Conjugate PE
Dilution Western Blot, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias Alternative macrophage activation-associated CC chemokine 1, AMAC1CC chemokine ligand 18, AMAC-1Small-inducible cytokine A18, chemokine (C-C motif) ligand 18 (pulmonary and activation-regulated), CKb7, DC-CK1CC chemokine PARC, DCCK1Macrophage inflammatory protein 4, Dendritic cell chemokine 1, MIP4, MIP-4C-C motif chemokine 18, PARCPulmonary and activation-regulated chemokine, SCYA18chemokine (C-C), dendritic, small inducible cytokine A18, small inducible cytokine subfamily A (Cys-Cys), member 18, pulmonary andactivation-regulated
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL18/PARC, Ala21-Ala89, Accession # P55774.1
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6362
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.