Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL19/MIP-3 beta Antibody (OTI2A12), HRP, Novus Biologicals™
SDP

Catalog No. NBP271278H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271278H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271278H Supplier Novus Biologicals Supplier No. NBP271278H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCL19/MIP-3 beta Monoclonal specifically detects CCL19/MIP-3 beta in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen CCL19/MIP-3 beta
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI2A12
Conjugate HRP
Dilution Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias beta chemokine exodus-3, Beta-chemokine exodus-3, CC chemokine ligand 19, C-C motif chemokine 19, chemokine (C-C motif) ligand 19, CKb11, EBI1-ligand chemokine, ELCMIP-3-beta, Epstein-Barr virus-induced molecule 1 ligand chemokine, exodus-3, Macrophage inflammatory protein 3 beta, macrophage inflammatory protein 3-beta, MGC34433, MIP-3b, MIP3BCK beta-11, SCYA19EBI1 ligand chemokine, small inducible cytokine subfamily A (Cys-Cys), member 19, Small-inducible cytokine A19
Gene Symbols CCL19
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 22-98 of human CCL19/MIP-3 beta (NP_006265) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 6363.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.