Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL2/JE/MCP-1 Antibody (001), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB074842 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB074842 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB074842 Supplier Novus Biologicals Supplier No. NBP290550AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen CCL2/JE/MCP-1
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, Mcp-1, SCYA2, SMC-CF
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CCL2/JE/MCP-1 (NP_035463.1; Met1-Asn148).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6347
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.