Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL2/JE/MCP-1 Antibody (123616R), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB312262 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312262 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312262 Supplier Novus Biologicals Supplier No. FAB479RPECY55
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CCL2/JE/MCP-1
Applications Flow Cytometry
Classification Monoclonal
Clone 123616R
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, Mcp-1, SCYA2, SMC-CF
Host Species Rat
Immunogen E. coli-derived recombinant mouse CCL2, Gln24-Arg96, Accession # P10148
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6347
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.