Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL2/JE/MCP-1 Antibody (24822), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB304272 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304272 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304272 Supplier Novus Biologicals Supplier No. FAB279A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CCL2/JE/MCP-1 Monoclonal antibody specifically detects CCL2/JE/MCP-1 in Human samples. It is validated for Western Blot, Neutralization

Specifications

Antigen CCL2/JE/MCP-1
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 24822
Conjugate APC
Dilution Western Blot, Neutralization
Formulation PBS
Gene Alias C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, Mcp-1, SCYA2, SMC-CF
Host Species Mouse
Immunogen E. coli-derived recombinant human CCL2/JE/MCP-1
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6347
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.