Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCL2/MCP1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB201929 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB201929 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB201929 Supplier Novus Biologicals Supplier No. NBP241209AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCL2/MCP1 Polyclonal antibody specifically detects CCL2/MCP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen CCL2/MCP1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias C-C motif chemokine 2, chemokine (C-C motif) ligand 2, GDCF-2, HC11Monocyte secretory protein JE, MCAFMonocyte chemotactic and activating factor, MCP1 Monocyte chemotactic protein 1, MCP-1 Small-inducible cytokine A2, MGC9434, Monocyte chemoattractant protein 1, monocyte chemoattractant protein-1, SCYA2HSMCR30, small inducible cytokine A2 (monocyte chemotactic protein 1, homologous tomouse Sig-je), small inducible cytokine subfamily A (Cys-Cys), member 2, SMC-CF
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid peptide near the carboxy terminus of human CCL2. The immunogen is located within the last 50 amino acids of CCL2. Amino Acid Squence: ADPKQKWVQDSMDHLD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Asthma, Biologically Active Proteins, Cancer, Chemokines and Cytokines, Cytokine Research, Hypoxia, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6347
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.